Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Plasmalemma vesicle-associated protein (PLVAP) | 50UG
SDP

Catalog No. 5000177573
Encompass_Preferred

Recombinant human plasmalemma vesicle-associated protein (PLVAP) fragment (Tyr49-Gly442) expressed in HEK293 cells with an N-terminal His tag. Supplied lyophilized from a PBS-based buffer containing 200 mM L-arginine and 8% trehalose, the protein is suitable for research into endothelial fenestrae, vascular permeability, and PLVAP interactions. Store lyophilized at -20 °C; after reconstitution aliquot and freeze for long-term storage.

  • Human-derived HEK293 expression system.
  • N-terminal His tag for purification and detection.
  • Sequence corresponds to Tyr49-Gly442 of PLVAP.
  • Accession number Q9BX97 for sequence reference.
  • Apparent molecular weight ~50-70 kDa.
  • Lyophilized formulation with L-arginine and trehalose for stability.
  • Store lyophilized at -20 °C; aliquot and freeze after reconstitution.
  • Intended for research use in studies of endothelial biology.

Catalog No. 50-001-77573 Supplier Medchemexpress LLC Supplier No. HYP70087950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human plasmalemma vesicle-associated protein (PLVAP) fragment (Tyr49-Gly442) expressed in HEK293 cells with an N-terminal His tag. Supplied lyophilized from a PBS-based buffer containing 200 mM L-arginine and 8% trehalose, the protein is suitable for research into endothelial fenestrae, vascular permeability, and PLVAP interactions. Store lyophilized at -20 °C; after reconstitution aliquot and freeze for long-term storage.

  • Human-derived HEK293 expression system.
  • N-terminal His tag for purification and detection.
  • Sequence corresponds to Tyr49-Gly442 of PLVAP.
  • Accession number Q9BX97 for sequence reference.
  • Apparent molecular weight ~50-70 kDa.
  • Lyophilized formulation with L-arginine and trehalose for stability.
  • Store lyophilized at -20 °C; aliquot and freeze after reconstitution.
  • Intended for research use in studies of endothelial biology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.