Learn More
Miele, Inc. PLW 8505 AC 115V undercounter labwasher
Catalog No. NC1828955
Encompass_Preferred
Simple and efficient, the PLW 8505 undercounter labwasher offers highly dependable and reliable performance. The modern design and unsurpassed technology with simple user interface enable mission-cricital uptime and cleaning for each laboratory and classroom space.
• 115V plug and play
• Intelligent controls: 9-program selection
• Visible 3.5" display
• All glass bottom-hinged door
• Electric heating
• Steam condenser
• Two integrated dispenser pumps
• Hot, cold and demineralized water
Catalog No. NC1828955 Supplier Miele, Inc. Supplier No. 11093230
Add to Cart
Add to Cart
Request a Quote
Simple and efficient, the PLW 8505 undercounter labwasher offers highly dependable and reliable performance. The modern design and unsurpassed technology with simple user interface enable mission-cricital uptime and cleaning for each laboratory and classroom space.
• 115V plug and play
• Intelligent controls: 9-program selection
• Visible 3.5" display
• All glass bottom-hinged door
• Electric heating
• Steam condenser
• Two integrated dispenser pumps
• Hot, cold and demineralized water