Learn More
Miele, Inc. PLW 8615 EL/S CT 3AC 208V large chamber labwasher
Catalog No. NC2738250
Encompass
With a smart variable-speed pump system andflexible chamber design featuring the SimpleLoad rackingsystem, Miele's PLW 86 series offers more glasswarererpocessing capacity. Certified with ACT LAbel
• 60 Bottles (1000mL)
• Up to 324 Bottles (100mL)
• 294 Vials
• 588 Pipettes
• SimpleLoad modules
• Rear basket docking
• 2-pump system: variable speed
• Auto liquid dispensing
• Multi-stage filtration system
• Spray arm monitoring
• PLW 8615 Combi Tank
Optional Features
• Drain Pump
• Rolling Plinth
• Drying unit - hot air HEPA drying
• Steam or Electric, or both
• Automatic or manual transfer cart
• I/OQ Validation
Catalog No. NC2738250 Supplier Miele, Inc. Supplier No. 11179560
Add to Cart
Add to Cart
Request a Quote
With a smart variable-speed pump system andflexible chamber design featuring the SimpleLoad rackingsystem, Miele's PLW 86 series offers more glasswarererpocessing capacity. Certified with ACT LAbel
• 60 Bottles (1000mL)
• Up to 324 Bottles (100mL)
• 294 Vials
• 588 Pipettes
• SimpleLoad modules
• Rear basket docking
• 2-pump system: variable speed
• Auto liquid dispensing
• Multi-stage filtration system
• Spray arm monitoring
• PLW 8615 Combi Tank
Optional Features
• Drain Pump
• Rolling Plinth
• Drying unit - hot air HEPA drying
• Steam or Electric, or both
• Automatic or manual transfer cart
• I/OQ Validation