Learn More
Miele, Inc. PLW 8617 EL/S TH 3AC 208V large chamber labwasher
Catalog No. NC2738557
Encompass
With a smart variable-speed pump system andflexible chamber design featuring the SimpleLoad rackingsystem, Miele's PLW 86 series offers more glasswarererpocessing capacity.
• 60 Bottles (1000mL)
• Up to 324 Bottles (100mL)
• 294 Vials
• 588 Pipettes
• SimpleLoad modules
• Rear basket docking
• 2-pump system: variable speed
• Auto liquid dispensing
• Multi-stage filtration system
• Spray arm monitoring
• PLW 8615 Combi TankOptional Features• Drain Pump• Rolling Plinth• Drying unit - hot air HEPA drying• Steam or Electric, or both• Automatic or manual transfer cart• I/OQ Validation
Catalog No. NC2738557 Supplier Miele, Inc. Supplier No. 11179830
Add to Cart
Add to Cart
Request a Quote
With a smart variable-speed pump system andflexible chamber design featuring the SimpleLoad rackingsystem, Miele's PLW 86 series offers more glasswarererpocessing capacity.
• 60 Bottles (1000mL)
• Up to 324 Bottles (100mL)
• 294 Vials
• 588 Pipettes
• SimpleLoad modules
• Rear basket docking
• 2-pump system: variable speed
• Auto liquid dispensing
• Multi-stage filtration system
• Spray arm monitoring
• PLW 8615 Combi TankOptional Features• Drain Pump• Rolling Plinth• Drying unit - hot air HEPA drying• Steam or Electric, or both• Automatic or manual transfer cart• I/OQ Validation