Learn More
Miele, Inc. PLW 6111 3AC 208V free standing labwasher
Catalog No. 501953622
Encompass_Preferred
Compact dimension, high capacity. Miele's SlimLine labwasher PLW 6111 offers a comprehensive and systematic solution toreprocessing laboratory glassware for analytical experiments.
• Slim design, 25 9/16 in.
(650mm) wide• SmartLoad 3 Racks,4 Levels
• Up to 126 narrow-neckedglasses or 121 pipettes
• 50L carboy
• 3.5" display withintelligent controls
• Rear basket docking
• Steam condenser
• Auto liquid dispensing
• Variable speed pump
• Multi-stage filtration system
• Spray arm monitoring
• DryPlus - hot air HEPA drying
Catalog No. 50-195-3622 Supplier Miele, Inc. Supplier No. 10959290
Add to Cart
Add to Cart
Request a Quote
Compact dimension, high capacity. Miele's SlimLine labwasher PLW 6111 offers a comprehensive and systematic solution toreprocessing laboratory glassware for analytical experiments.
• Slim design, 25 9/16 in.
(650mm) wide• SmartLoad 3 Racks,4 Levels
• Up to 126 narrow-neckedglasses or 121 pipettes
• 50L carboy
• 3.5" display withintelligent controls
• Rear basket docking
• Steam condenser
• Auto liquid dispensing
• Variable speed pump
• Multi-stage filtration system
• Spray arm monitoring
• DryPlus - hot air HEPA drying