Learn More
Apexbio Technology LLC PLX-4720, 100mg. Cas: 918505-84-7 MFCD: MFCD14635203. BRAF kinase inhibitor
SDP
Supplier:  Apexbio Technology LLC A3016100
Encompass_Preferred
PLX-4720, a 7-azaindole derivative discovered by a structure-guided discovery approach, is a potent inhibitor of B-RafV600E, the most frequent oncogenic protein kinase mutation, with the value of 50% inhibition concentration IC50 of 13 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4781