Learn More
Medchemexpress LLC HY-51424 10mg Medchemexpress, PLX-4720 CAS:918505-84-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5142410MG
Encompass_Preferred
Medchemexpress, HY-51424 10mg PLX-4720 CAS:918505-84-7 PLX-4720 is a potent and selective inhibitor of B-RafV600E with IC50 of 13 nM in a cell-free assay, equally potent to c-Raf-1(Y340D and Y341D mutations), and 10-fold selectivity for B-RafV600E than wild-type B-Raf. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9359
 Restricted Item