Learn More
Apexbio Technology LLC PLX-4720 918505-84-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064111
Encompass_Preferred
PLX-4720 (CAS 918505-84-7) is a selective inhibitor of the oncogenic kinase B-RafV600E a frequently observed activating mutation in cancer Structurally derived from a 7-azaindole scaffold PLX-4720 inhibits B-RafV600E with an IC50 of 13 nM exhibiting marked selectivity in comparison to wild-type B-Raf (IC50 160 nM) and other kinases such as FRK CSK SRC FAK FGFR and Aurora A (IC50 1000 nM) PLX-4720 decreases ERK phosphorylation in B-RafV600E-containing cells induces growth arrest and apoptosis in B-RafV600E melanoma lines and delays tumor progression in xenograft models harboring this mutation It represents a valuable tool in cancer research targeting B-Raf signaling pathways
Catalog No. 50-000-64111 Supplier Apexbio Technology LLC Supplier No. A30165.1
PLX-4720 (CAS 918505-84-7) is a selective inhibitor of the oncogenic kinase B-RafV600E a frequently observed activating mutation in cancer Structurally derived from a 7-azaindole scaffold PLX-4720 inhibits B-RafV600E with an IC50 of 13 nM exhibiting marked selectivity in comparison to wild-type B-Raf (IC50 160 nM) and other kinases such as FRK CSK SRC FAK FGFR and Aurora A (IC50 1000 nM) PLX-4720 decreases ERK phosphorylation in B-RafV600E-containing cells induces growth arrest and apoptosis in B-RafV600E melanoma lines and delays tumor progression in xenograft models harboring this mutation It represents a valuable tool in cancer research targeting B-Raf signaling pathways