Learn More
Selleck Chemical LLC PLX-4720 25mg 918505-84-7
SDP
Catalog No. 508445
Encompass_Preferred
PLX4720 is a potent and selective inhibitor of B-RafV600E with IC50 of 13 nM in a cell-free assay, equally potent to c-Raf-1(Y340D and Y341D mutations), 10-fold selectivity for B-RafV600E than wild-type B-Raf. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-844-5 Supplier Selleck Chemical LLC Supplier No. S115225MG
PLX4720 is a potent and selective inhibitor of B-RafV600E with IC50 of 13 nM in a cell-free assay, equally potent to c-Raf-1(Y340D and Y341D mutations), 10-fold selectivity for B-RafV600E than wild-type B-Raf. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.