Learn More
AdipoGen PLX4720 (25 mg)
SDP
Supplier:  AdipoGen AGCR13577M025
Encompass_Preferred
Chemical. CAS: 918505-84-7. Formula: C17H14ClF2N3O3S. MW: 413.8. Synthetic. Potent and selective cell permeable B-Raf (V600E) and c-Raf-1 (Y340D/Y341D) inhibitor. Antitumor compound. ERK phosphorylation is potently inhibited exclusive in B-Raf(V600E)-bearing tumor cell lines.
Catalog No. 50-205-3943