Learn More
AdipoGen PLX4720
SDP
Catalog No. 502562278
Encompass_Preferred
Chemical. CAS 918505-84-7. Formula C17H14ClF2N3O3S. MW 413.8. Synthetic. Potent and selective cell permeable B-Raf V600E and c-Raf-1 Y340D/Y341D inhibitor. Antitumor compound. ERK phosphorylation is potently inhibited exclusive in B-RafV600E-bearing tumor cell lines.
Catalog No. 50-256-2278 Supplier AdipoGen Supplier No. AGCR13577M005
Chemical. CAS 918505-84-7. Formula C17H14ClF2N3O3S. MW 413.8. Synthetic. Potent and selective cell permeable B-Raf V600E and c-Raf-1 Y340D/Y341D inhibitor. Antitumor compound. ERK phosphorylation is potently inhibited exclusive in B-RafV600E-bearing tumor cell lines.