Learn More
Medchemexpress LLC HY-114153 25mg , PLX5622 CAS:1303420-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11415325MG
Encompass_Preferred
HY-114153 25mg PLX5622 CAS: 1303420-67-8 PLX5622 is a highly selective brain penetrant and orally active CSF1R inhibitor (IC50=0.016 µM; Ki=5.9 nM). PLX5622 allows for extended and specific microglial elimination, preceding and during pathology development. PLX5622 demonstrates desirable PK properties in varies animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9030