Learn More
Selleck Chemical LLC PLX5622 2mg
SDP
Supplier:  Selleck Chemical LLC S88742MG
Encompass_Preferred
PLX5622 is a highly selective CSF-1R inhibitor (IC50 < 10 nmol/L), showing?>?20-fold selectivity over KIT and FLT3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3412