Learn More
Medchemexpress LLC HY-114153A 5mg Medchemexpress, PLX5622 (hemifumarate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114153A5MG
Encompass_Preferred
Medchemexpress, HY-114153A 5mg PLX5622 (hemifumarate) CAS: PLX5622 hemifumarate is a highly selective brain penetrant and oral active CSF1R inhibitor with an Ki of 5.9 nM, for extended and specific microglial elimination, preceding and during pathology development. PLX5622 hemifumarate demonstrates desirable PK properties in varies animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9292