Learn More
Selleck Chemical LLC PLX7904 5mg 1393465-84-3 PB04
SDP
Supplier:  Selleck Chemical LLC S79645MG
Encompass_Preferred
PLX7904, also known as PB04, is a potent and selective paradox-breaker RAF inhibitor. It is able to efficiently inhibit activation of ERK1/2 in mutant BRAF melanoma cells but does not hyperactivate ERK1/2 in mutant RAS-expressing cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5486