Learn More
Selleck Chemical LLC PLX8394 5mg 1393466-87-9
SDP
Catalog No. 501948184
Encompass_Preferred
PLX8394 is a next-generation, orally available, small-molecule BRAF inhibitor with IC50 values of 3.8 nM, 14 nM and 23 nM for BRAF(V600E), WT BRAF and CRAF respectively. It has potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8184 Supplier Selleck Chemical LLC Supplier No. S79655MG
PLX8394 is a next-generation, orally available, small-molecule BRAF inhibitor with IC50 values of 3.8 nM, 14 nM and 23 nM for BRAF(V600E), WT BRAF and CRAF respectively. It has potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.