Learn More
Aviva Systems Biology Corp Plxdc2 Antibody - C-terminal region (ARP50128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Plxdc2. It was validated on Western Blot. . Alternate names: 1200007L24Rik, 5430431D22Rik, AI646728, AU022916, AV231634, Tem7rApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-492