Learn More
Aviva Systems Biology Corp PLXNA4 Antibody - middle region (ARP58514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLXNA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLXNA4. Alternate names: DKFZp434G0625, DKFZp434G0625PRO34003, DKFZp566O0546, FAYV2820, FLJ35026, FLJ38287, KIAA1550, PLEXA4, PLXNA4A, PLXNA4B, PRO34003ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-62335