Learn More
Polysciences, Inc. Polybead Microspheres 0.10um, 15 mL
SDP
Catalog No. NC9081186
Encompass
Polybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90um. Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.
Catalog No. NC9081186 Supplier Polysciences, Inc. Supplier No. 0087615
Polybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90um. Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.