Learn More
Vector Laboratories Polyclonal Rabbit anti Human HAS2 Antibody (FITC), 200uL
SDP
Supplier:  Vector Laboratories LSC236105200
Encompass
HAS2 antibody LS-C236105 is a PE-conjugated rabbit polyclonal antibody to human HAS2 (aa145-174). Validated for ELISA, Flow, IHC and WB
Catalog No. NC2863576