Learn More
SETON SCIENTIFIC SETON SCIENTIFIC Polyclear Tubes, Open-Top, High Transparency, Puncturable, 50 Pack
Catalog No. NC9836096
Encompass
SETON SCIENTIFIC Polyclear Tubes are open-top, thin-wall centrifugation tubes ideal for scientific applications requiring high transparency. Designed from Polyclear thermoplastic, they withstand temperatures up to 25 °C and enable slice or puncture capability. Notably, they offer great visibility and are sterilizable by methods like ethylene oxide or 70% ethanol. With a nominal capacity of 12.9 mL and packaged in quantities of 50, these tubes differentiate from others by their optimal use in Cesium Chloride gradients without compromising performance.
Catalog No. NC9836096 Supplier SETON SCIENTIFIC Supplier No. 7031
SETON SCIENTIFIC Polyclear Tubes are open-top, thin-wall centrifugation tubes ideal for scientific applications requiring high transparency. Designed from Polyclear thermoplastic, they withstand temperatures up to 25 °C and enable slice or puncture capability. Notably, they offer great visibility and are sterilizable by methods like ethylene oxide or 70% ethanol. With a nominal capacity of 12.9 mL and packaged in quantities of 50, these tubes differentiate from others by their optimal use in Cesium Chloride gradients without compromising performance.