Learn More
Polylc Inc PolyLC PolyHYDROXYETHYL A Column, Exceptional Separation of Polar Compounds, HILIC Performance, 3.5 x 2.1 mm
SDP
Supplier:  Polylc Inc 3.52HY05006
Encompass
PolyLC PolyHYDROXYETHYL A column is a high-performance HPLC tool designed for Hydrophilic Interaction Chromatography (HILIC). This advanced column excels in separating polar compounds, providing sharper peaks and enhanced recoveries compared to competitors. Particularly effective for analyzing polar metabolites and proteins, it operates efficiently with high organic solvent concentrations, making it an indispensable choice for complex analytical tasks.
Catalog No. NC2606174