Learn More
AVANTOR MASTERFLEX (VWR LLC) Polyimide tubing, 0.0340""ID x 0.0380""OD, five 12""L/pack
Catalog No. NC1066026
Encompass
Polyimide tubing, 0.0340"ID x 0.0380"OD, five 12"L/pack, Applications: Use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers.Characteristics: Excellent chemical, heat, radiation, and abrasion resistance; high dielectric strength. Clear amber.Certification: Resin meets USP Class VI requirements and NEMA standards.Temperature range: ¿418 to 896degF (¿250 to 480degC)Sterilize: With autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC1066026 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9582007
Polyimide tubing, 0.0340"ID x 0.0380"OD, five 12"L/pack, Applications: Use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers.Characteristics: Excellent chemical, heat, radiation, and abrasion resistance; high dielectric strength. Clear amber.Certification: Resin meets USP Class VI requirements and NEMA standards.Temperature range: ¿418 to 896degF (¿250 to 480degC)Sterilize: With autoclave, ethylene oxide, or gamma irradiation.