Learn More
Perkin Elmer US LLC Brownlee Polypore H Cartridge Column - 220 mm x 4.6 mm I.D.
Catalog No. NC1882301
Encompass_Preferred
Suitable for the analysis of sugars and organic acids.
Catalog No. NC1882301 Supplier Perkin Elmer US LLC Supplier No. 07110089
Suitable for the analysis of sugars and organic acids.