Learn More
Perkin Elmer US LLC 10 mm Pre-Assembled Black Polypropylene Screw Cap with Pre-Slit PTFE/Silicone Septa, 100/pk
Catalog No. 509052363
Encompass_Preferred
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
Catalog No. 50-905-2363 Supplier Perkin Elmer US LLC Supplier No. N9306052
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.