Learn More
Ancare Corporation PLYSULFNE CAGE BTM, MOUSE20/CS
SDP
Catalog No. NC2217577
Encompass
Polysulfone Cage Bottom, Mouse, 11.5" x 7.5" x 5"HSlightly Tinted, Autoclavable to 270ºF (131ºC), 20 per case
This item is not returnable. View return policy
Catalog No. NC2217577 Supplier Ancare Corporation Supplier No. N10PLF
Polysulfone Cage Bottom, Mouse, 11.5" x 7.5" x 5"HSlightly Tinted, Autoclavable to 270ºF (131ºC), 20 per case