Learn More
Medchemexpress LLC PM-81I | 1637532-83-2 | 99.9% | 845.91 | 1 MG
SDP
Catalog No. 5000359312
Encompass_Preferred
PM-81I is a potent STAT6 inhibitor that targets the SH2 structural domain, effectively reducing STAT6 phosphorylation levels. This compound is valuable for studies in areas such as allergic lung disease, allergic rhinitis, chronic obstructive pulmonary disease, and cancer. It is supplied as a white to off-white solid with high purity.
  • Potent STAT6 inhibitor
  • Targets SH2 structural domain
  • Reduces STAT6 phosphorylation levels
  • Useful for allergic lung disease research
  • Useful for allergic rhinitis research
  • Useful for chronic obstructive pulmonary disease research
  • Useful for cancer research
  • White to off-white solid appearance
  • High purity
Catalog No. 50-003-59312 Supplier Medchemexpress LLC Supplier No. HY1480931MG
PM-81I is a potent STAT6 inhibitor that targets the SH2 structural domain, effectively reducing STAT6 phosphorylation levels. This compound is valuable for studies in areas such as allergic lung disease, allergic rhinitis, chronic obstructive pulmonary disease, and cancer. It is supplied as a white to off-white solid with high purity.
  • Potent STAT6 inhibitor
  • Targets SH2 structural domain
  • Reduces STAT6 phosphorylation levels
  • Useful for allergic lung disease research
  • Useful for allergic rhinitis research
  • Useful for chronic obstructive pulmonary disease research
  • Useful for cancer research
  • White to off-white solid appearance
  • High purity