Learn More
Caron Products And Services PM Kit Model 7000-33 Environmental Chamber
Catalog No. NC1784967
Encompass_Preferred
Preventative Maintenance Kit for Model 7000-33 (33 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (2) NOZ-110 Atomizers, Brass with Mesh Screen. Detailed Instructions Included.
Catalog No. NC1784967 Supplier Caron Products And Services Supplier No. PM700033
Preventative Maintenance Kit for Model 7000-33 (33 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (2) NOZ-110 Atomizers, Brass with Mesh Screen. Detailed Instructions Included.