Learn More
Chemglass Life Sciences Preventative Maintenance Kit for Overhead Drive Adapter
SDP
Catalog No. 50143172
Encompass
CGLS PM Kits include all of the high maintenance items required to keep your Overhead Drive Adapters functioning to the high standards.Each kit contains an illustrated parts list for easy identification of parts.Kit Includes: 1 each: #118 Silicone O-Ring 2 each: Shaft Seal 2 each: Polyester Ball Bearing 2 each: Internal Retaining Ring 1 each: Drive Shaft Grip Ring 1 each: Replacement Shaft
Catalog No. 50-143-172 Supplier Chemglass Life Sciences Supplier No. CLS1440K1
CGLS PM Kits include all of the high maintenance items required to keep your Overhead Drive Adapters functioning to the high standards.Each kit contains an illustrated parts list for easy identification of parts.Kit Includes: 1 each: #118 Silicone O-Ring 2 each: Shaft Seal 2 each: Polyester Ball Bearing 2 each: Internal Retaining Ring 1 each: Drive Shaft Grip Ring 1 each: Replacement Shaft