Learn More
Caron Products And Services PM Kit Model 7000-10 Environmental Chamber
Catalog No. NC1978126
Encompass_Preferred
Preventative Maintenance Kit for Model 7000-10 (10 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (1) NOZ-110 Atomizer, Brass with Mesh Screen. Detailed Instructions Included.
Catalog No. NC1978126 Supplier Caron Products And Services Supplier No. PM700010
Preventative Maintenance Kit for Model 7000-10 (10 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (1) NOZ-110 Atomizer, Brass with Mesh Screen. Detailed Instructions Included.