Learn More
Caron Products And Services PM Kit Model 7000-25 Environmental Chamber
Catalog No. NC2133306
Encompass_Preferred
Preventative Maintenance Kit for Model 7000-25 (25 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (2) NOZ-110 Atomizers, Brass with Mesh Screen. Detailed Instructions Included.
Catalog No. NC2133306 Supplier Caron Products And Services Supplier No. PM700025
Preventative Maintenance Kit for Model 7000-25 (25 cu. ft. Environmental Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, and (2) NOZ-110 Atomizers, Brass with Mesh Screen. Detailed Instructions Included.