Learn More
Surmodics Inc PM/Scl 75, 50 µg
SDP
Supplier:  Surmodics Inc A17000
Encompass
The polymyositis/scleroderma (PM/Scl) complex is the human ortholog of the Saccharomyces cerevisiae exosome, which is involved in the processing and degradation of RNAs. PM/Scl 75 is one of the human subunits found in the nucleoplasm and, at higher concentrations, in the nucleolus. DIARECT™ antigens are for further manufacturing or research use only.
Catalog No. NC1820500