Learn More
Pyramex™ PM1010 Ear Muff
Catalog No. 19155486 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Removable strap, PM1010
Catalog No.
19-155-486 Removable strap, PM1010
1 options
Catalog No. 19-155-486 Supplier Pyramex™ Supplier No. PM1010
PM1010 Ear Muff is electrically insulated and features a removable secure fit strap
  • Large soft foam ear cups
  • Dielectric
  • Durable padded nylon headband
  • Removable secure fit strap holds ear muff in place when wearing hard hat
Specifications
Color Black, Green
ANSI S3.1974 Yes
Cushion Material Polyurethane
Headband Material ABS
Model PM1010
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type Passive Ear Muffs
Dielectric Yes
Style Over-the-head