Learn More
Pyramex™ PM2010 Series Passive Ear Muff
Catalog No. 19155487 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
22 dB
Catalog No. Noise Reduction Rating (NRR)
19-155-487 22 dB
1 options
Catalog No. 19-155-487 Supplier Pyramex™ Supplier No. PM2010
Add to Cart
Add to Cart
PM2010 Ear Muff features height adjustability to fit most head sizes.
  • Lightweight and comfortable
  • Dielectric
  • Durable padded nylon headband
  • Headband allows easy adjustment
Specifications
Color Black, Gray
ANSI S3.1974 Yes
Cushion Material PVC
Cushion Color Black
Headband Material Polycarbonate, ABS
Model PM2010
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 22 dB
Product Type Passive Ear Muffs
Headband Color Black
Dielectric Yes
Style Over-the-head