Learn More
Aviva Systems Biology Corp PM20D2 Antibody - middle region (ARP51811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PM20D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PM20D2. Alternate names: ACY1L2, bA63L7.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60622