Learn More
Caron Products And Services PM Kit Model 7401-33 Shaker Ready CO2 Incubator
Catalog No. NC1779535
Encompass_Preferred
Preventative Maintenance Kit for Model 7401-33 (33 cu. ft. Shaker Ready CO2 Incubator) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing and (1) FIL-213 Filter, .2 Micron.  Detailed Instructions Included.
Catalog No. NC1779535 Supplier Caron Products And Services Supplier No. PM740133
Preventative Maintenance Kit for Model 7401-33 (33 cu. ft. Shaker Ready CO2 Incubator) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing and (1) FIL-213 Filter, .2 Micron.  Detailed Instructions Included.