Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ PM80 Series Ear Muff

Catalog No. 19155491 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black, Gray
High-visibility Lime
High-visibility Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-491 High-visibility Orange
19-155-489 Black, Gray
19-155-490 High-visibility Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-155-491 Supplier Pyramex™ Supplier No. PM8041
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for a variety of industrial uses, the PM80 Ear Muff is durable, comfortable and features easy adjustability.

  • Durable material with excellent shape memory
  • Soft and comfortable
  • Easy to adjust, fits different head sizes
  • Fold-away headband

Specifications

Color High-visibility Orange
ANSI S3.1974 Yes
Cushion Material PVC
Cushion Color Black
Headband Material POM
For Use With (Application) Variety of industrial applications
Model PM8041
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Headband Color Black
Style Over-the-head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.