Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ PM90 Series Ear Muffs

Catalog No. 19155492 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black, Gray
Black, Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-492 Black, Gray
19-155-493 Black, Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-155-492 Supplier Pyramex™ Supplier No. PM9010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PM90 Ear Muffs low-profile design are constructed of durable material and feature excellent shape memory.

  • Durable material with excellent shape memory
  • Soft and comfortable
  • Easy to adjust, fits different head sizes
  • Ear cup and volume ring blocks out noise
  • Available in black or pink

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Material PVC
Cushion Color Black
Headband Material POM
For Use With (Application) Construction, Energy and Utilities, General Manufacturing, Mining, Oil and Gas, Transportation
Model PM9010
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Headband Color Black
Style Over-the-head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.