Learn More
Apexbio Technology LLC 12-O-tetradecanoyl phorbol-13-acetate (PMA),5mgCAS# 16561-29-8
SDP
Catalog No. 502239962
Encompass_Preferred
12-O-tetradecanoyl phorbol-13-acetate (TPA) is an activator of ERK/MAPK with the concentration of 50μm [1].ERK is an extracellular signal-regulated kinase and transfers signals from a receptor on the cell surface to DNA cooperated with MAPK. It is reported that ERK deficiency leads to the cell uncontrolled growth and is regarded as a target to cure cancers.TPA is a potent ERK activator. When tested with A549 cells (human lung cancer cell line), TPA treatment led to an early, strong, and relatively transient ERK phosphorylation [2]. In mouse embryo fibroblasts from DUSP5 (+/+) mice, administration of TPA increased levels of ERK expression [3]. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-223-9962 Supplier Apexbio Technology LLC Supplier No. N20605
12-O-tetradecanoyl phorbol-13-acetate (TPA) is an activator of ERK/MAPK with the concentration of 50μm [1].ERK is an extracellular signal-regulated kinase and transfers signals from a receptor on the cell surface to DNA cooperated with MAPK. It is reported that ERK deficiency leads to the cell uncontrolled growth and is regarded as a target to cure cancers.TPA is a potent ERK activator. When tested with A549 cells (human lung cancer cell line), TPA treatment led to an early, strong, and relatively transient ERK phosphorylation [2]. In mouse embryo fibroblasts from DUSP5 (+/+) mice, administration of TPA increased levels of ERK expression [3]. Other sizes are also available. Please inqury us for quote.