Learn More
Selleck Chemical LLC PMA
SDP
Catalog No. 5000001004
Encompass_Preferred
PMA (Phorbol 12-myristate 13-acetate) a potent activator of PKC is active at nanomolar concentrations Phorbol 12-myristate 13-acetate (PMA) induces sphingosine-1-phosphate (S1P) PMA has strong excitability on the skin and mucous membrane Special protection is required when using PMA Wear gloves and mask to avoid direct contact in any way
Catalog No. 50-000-01004 Supplier Selleck Chemical LLC Supplier No. S779110MM/1ML
Add to Cart
Add to Cart
PMA (Phorbol 12-myristate 13-acetate) a potent activator of PKC is active at nanomolar concentrations Phorbol 12-myristate 13-acetate (PMA) induces sphingosine-1-phosphate (S1P) PMA has strong excitability on the skin and mucous membrane Special protection is required when using PMA Wear gloves and mask to avoid direct contact in any way