Learn More
New England Biolabs, Inc. pMAL™-c6T Vector – 50 µl
SDP
Catalog No. 502024047
Encompass
The pMAL-c6T Vector is designed to produce maltose-binding protein (MBP) fusions in the cytoplasm. The MBP has been engineered for tighter binding to amylose resin. The vector expresses an N-terminal his-tagged malE gene followed by a multiple cloning site containing a TEV protease recognition sequence, allowing MBP to be cleaved from the protein of interest after purification.
Catalog No. 50-202-4047 Supplier New England Biolabs, Inc. Supplier No. N0378S
The pMAL-c6T Vector is designed to produce maltose-binding protein (MBP) fusions in the cytoplasm. The MBP has been engineered for tighter binding to amylose resin. The vector expresses an N-terminal his-tagged malE gene followed by a multiple cloning site containing a TEV protease recognition sequence, allowing MBP to be cleaved from the protein of interest after purification.