Learn More
Pendotech Pendotech Pressuremat Panel Mount Kit With Two Sensor Cables
Supplier:  Pendotech PMATPANEL2C
Encompass
PressureMAT panel mount kit - 2 gaskets, 4 mounting brakcets, 2 sensor cables, and input connectors for sensors and power. (Used with PMAT2, PMAT2HR, PMAT2A, PMAT2P, PMAT2F. Part number is used when PMAT was purchased separately.)
Catalog No. NC1831888