Learn More
Biotium PMAxx™, 20mM in H2O
Catalog No. NC1389321
Encompass_Preferred
PMAxx is a new and improved version of the viability dye PMA. It has higher activity than PMA, and thus a greater ability to differentiate between live and dead bacteria by qPCR.
Catalog No. NC1389321 Supplier Biotium Supplier No. 40069
PMAxx is a new and improved version of the viability dye PMA. It has higher activity than PMA, and thus a greater ability to differentiate between live and dead bacteria by qPCR.