Learn More
Biotium PMA Real-Time PCR Bacterial Viability Kit - Listeria monocytogenes (hly) PMAxx™
Catalog No. 501964566
Encompass_Preferred
This PMAxx™-PCR kit is designed for selective detection of viable bacteria from a specific strain using PMAxx™ dye and real-time PCR. This kit provides primers for specific amplification of the Listeria monocytogenes hly gene. The kit has sufficient reagents for treating of 80 bacterial cultures and performing 200 PCR reactions with PMAxx™. PMAxx™ is a new and improved version of PMA designed by Biotium scientists to be a superior alternative to PMA. While PMA is generally effective at differentiating between live and dead bacteria by qPCR, it does not completely eliminate PCR products from dead cell DNA. This could potentially give false positive results. Biotium’s new dye PMAxx™ is much more effective at eliminating PCR amplification of dead cell DNA, and therefore provides the best discrimination between live and dead bacteria. Listeria monocytogenes is a food-borne pathogen that can cause digestive illness and fetal harm.
Catalog No. 50-196-4566 Supplier Biotium Supplier No. 31051X
This PMAxx™-PCR kit is designed for selective detection of viable bacteria from a specific strain using PMAxx™ dye and real-time PCR. This kit provides primers for specific amplification of the Listeria monocytogenes hly gene. The kit has sufficient reagents for treating of 80 bacterial cultures and performing 200 PCR reactions with PMAxx™. PMAxx™ is a new and improved version of PMA designed by Biotium scientists to be a superior alternative to PMA. While PMA is generally effective at differentiating between live and dead bacteria by qPCR, it does not completely eliminate PCR products from dead cell DNA. This could potentially give false positive results. Biotium’s new dye PMAxx™ is much more effective at eliminating PCR amplification of dead cell DNA, and therefore provides the best discrimination between live and dead bacteria. Listeria monocytogenes is a food-borne pathogen that can cause digestive illness and fetal harm.