Learn More
Adam Equipment Adam Equipment PMB 163 Moisture Analyzer, with Halegon Lamp, 160g Capacity, 0.001g Readability
Supplier:  Adam Equipment PMB 163
Encompass_Preferred
Adam Equipment PMB moisture balances set a new standard for data communication making recording results faster and easier. Use the USB interface and connect a memory stick to download the results as they are taken and store them for future analysis. No need for additional software to take readings from the balance giving the user total freedom to collect data on a production floor or in the field. The PMB's fast response time and easy-to-use functionality make it the ideal moisture analyzer for a range of different applications. The automatic test setting function lets you quickly run multiple tests without additional user input and the built-in memories let you store that data future reference. The PMB offers smart features that have been developed to give you the ultimate performance for moisture analysis. Navigating the built in applications is easy with the intuitively designed keypad.
Catalog No. NC2021394