Learn More
Enzo Life Sciences PMCA ATPase monoclonal antibody (5F10) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804049R100
Encompass_Preferred
Clone: 5F10. Isotype: IgG2a. Immunogen: Purified human erythrocyte PMCA ATPase (plasma membrane calcium pump ATPase). Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Monkey, Dog, Chicken, Cat, Amphibian. Specificity: Recognizes all known (currently four) isoforms of plasma membrane PMCA ATPase. Recognizes an epitope between aa 724-783 of human PMCA ATPase. Applications: ICC, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9759