Learn More
Cenmed™ PMD Prosafe Disposable Premium Economy Lab Coats, White
SDP
Catalog No. 21520234
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-520-234 5X-Large
21-520-226 X-Small
21-520-227 Small
21-520-228 Medium
21-520-229 Large
21-520-230 X-Large
21-520-231 2X-Large
21-520-232 3X-Large
21-520-233 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-520-234 Supplier Cenmed™ Supplier No. B113660WH5XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Anti-static lab coats made of 3-layer SMS offer fluid resistance and are professional and comfortable.
  • Soft yet durable 3-ply SMS latex-free material
  • Fluid resistand and breathable for protective comfort
  • Knit cuffs and collar for a snug feel
  • Snap closure front, and 3-pockets
  • Anti-Static treate with no clinging
  • Washable (cold water) up to 5 times
  • Fire retardant
  • Tested tensile strength of 119.7 g/m2 MD and 61.6 g/m2 CD
  • Elongation testing showing 109 g/m2 MD and 118.9 g/m2 CD
  • Individually wrapped, sold in packs of 10
  • Knee length
Specifications
Product Type General Purpose Lab Coat
Size 5X-Large
Gender Unisex
Material SMS, Polypropylene
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Knee-length
No. of Pockets 3
Collar Type Knit
Finish Premium
Model B113600SB
Barrier Retention Hydrostatic Head of 38.2 cmH2O
Packaging Type Individually Packaged