Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ PMD Sterilization Trays with Pin Mat and Center Lid Retainer
SDP

Catalog No. 21520140
Change view
Click to view available options
Disposable:
Reusable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
21-520-140 Reusable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-140 Supplier Cenmed™ Supplier No. S20100002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Microsurgical Sterilization Case with pin mat retainers. Ideal for sterilizing a small set of instruments.

  • Contains pin mat and center lid retainer for secure storage
  • External dimensions: 5 x 10.5 x 2.5 in.
  • Internal dimensions: 4.50 x 10.29 x 1.53 in.

Specifications

Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) Sterilizers
Grade Premium
Product Line Sterilization Container, Sterilization Tray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.