Learn More
New England Biolabs, Inc. PmeI – 500 units
SDP
Catalog No. 50812245
Encompass
A restriction endonuclease that recognizes the sequence GTTT_^AAAC.
Catalog No. 50-812-245 Supplier New England Biolabs, Inc. Supplier No. R0560S
A restriction endonuclease that recognizes the sequence GTTT_^AAAC.