Learn More
Aviva Systems Biology Corp PMF1 Antibody - N-terminal region (ARP38922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PMF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PMF1. Alternate names: RP11-54H19.4ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53120